Anti-CEACAM1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 283340]

Supplier: R & D Systems
FAB2244A
220017-373EA 561.11 USD
220017-373
Anti-CEACAM1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 283340]
Antibodies
Type: Primary
Antigen: CEACAM1
Clonality: Monoclonal
Clone: 283340
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR