Anti-PVRL2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 610603]

Supplier: R & D Systems
FAB2229P
102881-914EA 480.12 USD
102881-914
Anti-PVRL2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 610603]
Antibodies
Type: Primary
Antigen: PVRL2
Clonality: Monoclonal
Clone: 610603
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR