Anti-NCR1 Goat Polyclonal Antibody (Fluorescein)

Supplier: R & D Systems
FAB2225F-100 FAB2225F-025
102881-900EA 177.88 USD
102881-900 102881-902
Anti-NCR1 Goat Polyclonal Antibody (Fluorescein)
Antibodies
Type: Primary
Antigen: NCR1
Clonality: Polyclonal
Clone:
Conjugation: Fluorescein
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR