Anti-NRP2 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 257103]

Supplier: R & D Systems
FAB22151A
102881-878EA 582.8 USD
102881-878
Anti-NRP2 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 257103]
Antibodies
Type: Primary
Antigen: NRP2
Clonality: Monoclonal
Clone: 257103
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR