Anti-CA9 Mouse Monoclonal Antibody (Fluorescein) [clone: 303123]

Supplier: R & D Systems
FAB2188F
220017-365EA 425.17 USD
220017-365
Anti-CA9 Mouse Monoclonal Antibody (Fluorescein) [clone: 303123]
Antibodies
Type: Primary
Antigen: CA9
Clonality: Monoclonal
Clone: 303123
Conjugation: Fluorescein
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR