Anti-CXCR4 Rat Monoclonal Antibody (Fluorescein) [clone: 247506]

Supplier: R & D Systems
FAB21651F-025 FAB21651F-100
102881-870EA 160.52 USD
102881-870 102881-872
Anti-CXCR4 Rat Monoclonal Antibody (Fluorescein) [clone: 247506]
Antibodies
Type: Primary
Antigen: CXCR4
Clonality: Monoclonal
Clone: 247506
Conjugation: Fluorescein
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR