Anti-CXCR2 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 242216]

Supplier: R & D Systems
FAB2164A-025 FAB2164A-100
102881-852EA 543.75 USD
102881-852 102881-850
Anti-CXCR2 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 242216]
Antibodies
Type: Primary
Antigen: CXCR2
Clonality: Monoclonal
Clone: 242216
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR