Anti-CCR9 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 242503]

Supplier: R & D Systems
FAB2160P-025 FAB2160P-100
102881-846EA 177.88 USD
102881-846 102881-848
Anti-CCR9 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 242503]
Antibodies
Type: Primary
Antigen: CCR9
Clonality: Monoclonal
Clone: 242503
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR