Anti-CCR9 Rat Monoclonal Antibody (Fluorescein) [clone: 242503]

Supplier: R & D Systems
FAB2160F-100 FAB2160F-025
102881-842EA 160.52 USD
102881-842 102881-844
Anti-CCR9 Rat Monoclonal Antibody (Fluorescein) [clone: 242503]
Antibodies
Type: Primary
Antigen: CCR9
Clonality: Monoclonal
Clone: 242503
Conjugation: Fluorescein
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR