Anti-VTCN1 Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: 297219]

Supplier: R & D Systems
FAB2154G
102881-826EA 543.75 USD
102881-826
Anti-VTCN1 Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: 297219]
Antibodies
Type: Primary
Antigen: VTCN1
Clonality: Monoclonal
Clone: 297219
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR