Anti-LDLR Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: 472413]

Supplier: R & D Systems
FAB2148N
102881-822EA 543.75 USD
102881-822
Anti-LDLR Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: 472413]
Antibodies
Type: Primary
Antigen: LDLR
Clonality: Monoclonal
Clone: 472413
Conjugation: Alexa Fluor® 700
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR