Anti-CXCR6 Rat Monoclonal Antibody (Alexa Fluor® 700) [clone: 221002]

Supplier: R & D Systems
FAB2145N-025 FAB2145N-100
102881-812EA 513.38 USD
102881-812 102881-810
Anti-CXCR6 Rat Monoclonal Antibody (Alexa Fluor® 700) [clone: 221002]
Antibodies
Type: Primary
Antigen: CXCR6
Clonality: Monoclonal
Clone: 221002
Conjugation: Alexa Fluor® 700
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR