Anti-CXCR6 Rat Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: 221002]

Supplier: R & D Systems
FAB2145C-025 FAB2145C-100
102881-802EA 202.46 USD
102881-802 102881-804
Anti-CXCR6 Rat Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: 221002]
Antibodies
Type: Primary
Antigen: CXCR6
Clonality: Monoclonal
Clone: 221002
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR