Anti-LYVE1 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 223322]

Supplier: R & D Systems
FAB2125P
102881-794EA 533.63 USD
102881-794
Anti-LYVE1 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 223322]
Antibodies
Type: Primary
Antigen: LYVE1
Clonality: Monoclonal
Clone: 223322
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR