Anti-LILRB2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 287219]

Supplier: R & D Systems
FAB2078P-100 FAB2078P-025
102881-736EA 507.6 USD
102881-736 102881-734
Anti-LILRB2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 287219]
Antibodies
Type: Primary
Antigen: LILRB2
Clonality: Monoclonal
Clone: 287219
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR