Anti-LILRB2 Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: 287219]

Supplier: R & D Systems
FAB2078N-025 FAB2078N-100
102881-732EA 513.38 USD
102881-732 102881-730
Anti-LILRB2 Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: 287219]
Antibodies
Type: Primary
Antigen: LILRB2
Clonality: Monoclonal
Clone: 287219
Conjugation: Alexa Fluor® 700
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR