Anti-FCGR1 Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: 290322]

Supplier: R & D Systems
FAB20741G
102881-722EA 485.91 USD
102881-722
Anti-FCGR1 Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: 290322]
Antibodies
Type: Primary
Antigen Symbol: FCGR1
Antigen Name: Fc receptor, IgG, high affinity I
Antigen Synonyms: AI323638, AV092959, CD64, FcgammaRI, IGGHAFC
Clonality: Monoclonal
Clone: 290322
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR