Anti-SEMA7A Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 310829]

Supplier: R & D Systems
FAB20681A
102881-716EA 584.25 USD
102881-716
Anti-SEMA7A Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 310829]
Antibodies
Type: Primary
Antigen: SEMA7A
Clonality: Monoclonal
Clone: 310829
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR