Anti-THY1 Mouse Monoclonal Antibody (PE) [clone: Thy-1A1]

Supplier: R & D Systems
FAB2067P
220017-330EA 419.38 USD
220017-330
Anti-THY1 Mouse Monoclonal Antibody (PE) [clone: Thy-1A1]
Antibodies
Type: Primary
Antigen Symbol: THY1
Antigen Name: Thy-1 cell surface antigen
Antigen Synonyms: CD90, CDw90
Clonality: Monoclonal
Clone: Thy-1A1
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR