Anti-THY1 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: Thy-1A1]

Supplier: R & D Systems
FAB2067G
102881-714EA 454.09 USD
102881-714
Anti-THY1 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: Thy-1A1]
Antibodies
Type: Primary
Antigen Symbol: THY1
Antigen Name: Thy-1 cell surface antigen
Antigen Synonyms: CD90, CDw90
Clonality: Monoclonal
Clone: Thy-1A1
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR