Anti-ROR2 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 231512]

Supplier: R & D Systems
FAB2064G
102881-712EA 561.11 USD
102881-712
Anti-ROR2 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 231512]
Antibodies
Type: Primary
Antigen: ROR2
Clonality: Monoclonal
Clone: 231512
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR