Anti-ROR2 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 231509]

Supplier: R & D Systems
FAB20641A
102881-704EA 608.83 USD
102881-704
Anti-ROR2 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 231509]
Antibodies
Type: Primary
Antigen: ROR2
Clonality: Monoclonal
Clone: 231509
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR