Anti-FCRL2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 296902]

Supplier: R & D Systems
FAB2048P
220017-325EA 513.38 USD
220017-325
Anti-FCRL2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 296902]
Antibodies
Type: Primary
Antigen Symbol: FCRL2
Antigen Name: Fc receptor-like 2
Antigen Synonyms: CD307b, FCRH2, IFGP4, IRTA4, SPAP1, SPAP1A, SPAP1B, SPAP1C
Clonality: Monoclonal
Clone: 296902
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR