Anti-SPN Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 290111]

Supplier: R & D Systems
FAB2038P
220017-321EA 389.02 USD
220017-321
Anti-SPN Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 290111]
Antibodies
Type: Primary
Antigen: SPN
Clonality: Monoclonal
Clone: 290111
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR