Anti-LILRB1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 292305]

Supplier: R & D Systems
FAB20171P-100 FAB20171P-025
102881-698EA 480.12 USD
102881-698 102881-696
Anti-LILRB1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 292305]
Antibodies
Type: Primary
Antigen: LILRB1
Clonality: Monoclonal
Clone: 292305
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR