Anti-LILRB1 Mouse Monoclonal Antibody (Fluorescein) [clone: 292305]

Supplier: R & D Systems
FAB20171F-025 FAB20171F-100
102881-694EA 389.02 USD
102881-694 102881-692
Anti-LILRB1 Mouse Monoclonal Antibody (Fluorescein) [clone: 292305]
Antibodies
Type: Primary
Antigen: LILRB1
Clonality: Monoclonal
Clone: 292305
Conjugation: Fluorescein
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR