Anti-S1PR1 Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: 218713]

Supplier: R & D Systems
FAB2016N
102881-686EA 543.75 USD
102881-686
Anti-S1PR1 Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: 218713]
Antibodies
Type: Primary
Antigen: S1PR1
Clonality: Monoclonal
Clone: 218713
Conjugation: Alexa Fluor® 700
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR