Anti-S1PR1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 218713]

Supplier: R & D Systems
FAB2016A
220017-315EA 594.37 USD
220017-315
Anti-S1PR1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 218713]
Antibodies
Type: Primary
Antigen: S1PR1
Clonality: Monoclonal
Clone: 218713
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR