Anti-KIR2DL3 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 180701]

Supplier: R & D Systems
FAB2014A-025 FAB2014A-100
102881-672EA 543.75 USD
102881-672 102881-462
Anti-KIR2DL3 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 180701]
Antibodies
Type: Primary
Antigen: KIR2DL3
Clonality: Monoclonal
Clone: 180701
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR