Anti-IL1A Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 3405]

Supplier: R & D Systems
FAB200P
102881-460EA 633.42 USD
102881-460
Anti-IL1A Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 3405]
Antibodies
Type: Primary
Antigen: IL1A
Clonality: Monoclonal
Clone: 3405
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR