Anti-ROR1 Goat Polyclonal Antibody (Alexa Fluor® 488)

Supplier: R & D Systems
FAB2000G
102881-452EA 561.11 USD
102881-452
Anti-ROR1 Goat Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
Type: Primary
Antigen: ROR1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR