Anti-ITGAE Goat Polyclonal Antibody (Alexa Fluor® 488)

Supplier: R & D Systems
FAB1990G
102881-662EA 323.94 USD
102881-662
Anti-ITGAE Goat Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
Type: Primary
Antigen: ITGAE
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR