Anti-FZD7 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 151143]

Supplier: R & D Systems
FAB1981A
220017-304EA 577.02 USD
220017-304
Anti-FZD7 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 151143]
Antibodies
Type: Primary
Antigen Symbol: FZD7
Antigen Name: frizzled homolog 7 (Drosophila)
Antigen Synonyms: Fz7
Clonality: Monoclonal
Clone: 151143
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR