Anti-CCR7 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 150503]

Supplier: R & D Systems
FAB197P-025 FAB197P-100
102881-660EA 484.46 USD
102881-660 102881-658
Anti-CCR7 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 150503]
Antibodies
Type: Primary
Antigen: CCR7
Clonality: Monoclonal
Clone: 150503
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR