Anti-CCR7 Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: 150503]

Supplier: R & D Systems
FAB197N-025 FAB197N-100
102881-654EA 160.52 USD
102881-654 102881-656
Anti-CCR7 Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: 150503]
Antibodies
Type: Primary
Antigen: CCR7
Clonality: Monoclonal
Clone: 150503
Conjugation: Alexa Fluor® 700
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR