Anti-CCR7 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 150503]

Supplier: R & D Systems
FAB197A-100 FAB197A-025
102881-640EA 221.26 USD
102881-640 102881-642
Anti-CCR7 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 150503]
Antibodies
Type: Primary
Antigen: CCR7
Clonality: Monoclonal
Clone: 150503
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR