Anti-FCGR3 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 275003]

Supplier: R & D Systems
FAB19601P-100 FAB19601P-025
102881-636EA 193.78 USD
102881-636 102881-638
Anti-FCGR3 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 275003]
Antibodies
Type: Primary
Antigen Symbol: FCGR3
Antigen Name: Fc receptor, IgG, low affinity III
Antigen Synonyms: CD16
Clonality: Monoclonal
Clone: 275003
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR