Anti-FCGR3 Rat Monoclonal Antibody (Fluorescein) [clone: 275003]

Supplier: R & D Systems
FAB19601F-025 FAB19601F-100
102881-630EA 339.85 USD
102881-630 102881-628
Anti-FCGR3 Rat Monoclonal Antibody (Fluorescein) [clone: 275003]
Antibodies
Type: Primary
Antigen Symbol: FCGR3
Antigen Name: Fc receptor, IgG, low affinity III
Antigen Synonyms: CD16
Clonality: Monoclonal
Clone: 275003
Conjugation: Fluorescein
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR