Anti-CCR6 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 53103]

Supplier: R & D Systems
FAB195P-100 FAB195P-025
102881-618EA 501.82 USD
102881-618 102881-616
Anti-CCR6 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 53103]
Antibodies
Type: Primary
Antigen: CCR6
Clonality: Monoclonal
Clone: 53103
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR