Anti-CCR6 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 53103]

Supplier: R & D Systems
FAB195A-025 FAB195A-100
102881-604EA 177.88 USD
102881-604 102881-606
Anti-CCR6 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 53103]
Antibodies
Type: Primary
Antigen: CCR6
Clonality: Monoclonal
Clone: 53103
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR