Anti-IL12RB2 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 305719]

Supplier: R & D Systems
FAB1959G-025 FAB1959G-100
102881-596EA 286.34 USD
102881-596 102881-598
Anti-IL12RB2 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 305719]
Antibodies
Type: Primary
Antigen: IL12RB2
Clonality: Monoclonal
Clone: 305719
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR