Anti-TNFRSF17 Goat Polyclonal Antibody (Fluorescein)

Supplier: R & D Systems
FAB193F
220017-282EA 438.18 USD
220017-282
Anti-TNFRSF17 Goat Polyclonal Antibody (Fluorescein)
Antibodies
Type: Primary
Antigen: TNFRSF17
Clonality: Polyclonal
Clone:
Conjugation: Fluorescein
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR