Anti-TNFRSF17 Goat Polyclonal Antibody (APC (Allophycocyanin))

Supplier: R & D Systems
FAB193A
220017-281EA 600.15 USD
220017-281
Anti-TNFRSF17 Goat Polyclonal Antibody (APC (Allophycocyanin))
Antibodies
Type: Primary
Antigen: TNFRSF17
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR