Anti-CXCR5 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 51505]

Supplier: R & D Systems
FAB190P-100 FAB190P-025
102881-584EA 582.8 USD
102881-584 102881-582
Anti-CXCR5 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 51505]
Antibodies
Type: Primary
Antigen: CXCR5
Clonality: Monoclonal
Clone: 51505
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR