Anti-CXCR5 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 51505]

Supplier: R & D Systems
FAB190A-025 FAB190A-100
102881-376EA 644.98 USD
102881-376 102881-374
Anti-CXCR5 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 51505]
Antibodies
Type: Primary
Antigen: CXCR5
Clonality: Monoclonal
Clone: 51505
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR