Anti-SLAMF6 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 292811]

Supplier: R & D Systems
FAB19081P-025 FAB19081P-100
102881-372EA 513.38 USD
102881-372 102881-370
Anti-SLAMF6 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 292811]
Antibodies
Type: Primary
Antigen: SLAMF6
Clonality: Monoclonal
Clone: 292811
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR