Anti-SLAMF6 Mouse Monoclonal Antibody (Fluorescein) [clone: 292811]

Supplier: R & D Systems
FAB19081F-100 FAB19081F-025
102881-368EA 404.92 USD
102881-368 102881-366
Anti-SLAMF6 Mouse Monoclonal Antibody (Fluorescein) [clone: 292811]
Antibodies
Type: Primary
Antigen: SLAMF6
Clonality: Monoclonal
Clone: 292811
Conjugation: Fluorescein
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR