Anti-SLAMF8 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 250014]

Supplier: R & D Systems
FAB19072P
220017-269EA 513.38 USD
220017-269
Anti-SLAMF8 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 250014]
Antibodies
Type: Primary
Antigen: SLAMF8
Clonality: Monoclonal
Clone: 250014
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR