Anti-SLAMF7 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 235614]

Supplier: R & D Systems
FAB1906A
220017-266EA 608.83 USD
220017-266
Anti-SLAMF7 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 235614]
Antibodies
Type: Primary
Antigen: SLAMF7
Clonality: Monoclonal
Clone: 235614
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR