Anti-KLRF1 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 239127]

Supplier: R & D Systems
FAB1900A-100 FAB1900A-025
102881-566EA 211.14 USD
102881-566 102881-568
Anti-KLRF1 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 239127]
Antibodies
Type: Primary
Antigen: KLRF1
Clonality: Monoclonal
Clone: 239127
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR